Assay Details
Target Gene Details
Entrez Gene ID: | 10380 |
Gene Name: | 3'(2'), 5'-bisphosphate nucleotidase 1 |
Gene Aliases: |
HEL20, PIP |
Location: |
Chr.1:220057482-220089916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832636 | Chr.1:219966355 - 220121010 on Build GRCh38 | Loss |
|
| nsv4632 | Chr.1:220041527 - 220076852 on Build GRCh38 | Insertion |
|
| nsv549222 | Chr.1:220053390 - 220073050 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map