Assay Details
Target Gene Details
Entrez Gene ID: | 1410 |
Gene Name: | crystallin alpha B |
Gene Aliases: |
CMD1II, CRYA2, CTPP2, CTRCT16, HEL-S-101, HSPB5, MFM2 |
Location: |
Chr.11:111908620-111913213 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYAB | NM_001289807.1 | 4 | 879 | NP_001276736.1 |
| NM_001289808.1 | 3 | 908 | NP_001276737.1 | |
| NM_001885.2 | 4 | 913 | NP_001876.1 | |
| XM_011542608.1 | 4 | 894 | XP_011540910.1 | |
| XM_011542609.2 | 3 | 1135 | XP_011540911.1 | |
| AF007162.1 | 3 | 635 | AAC19161.1 | |
| BC007008.1 | 3 | 652 | AAH07008.1 | |
| BF339748.1 | 1 | 187 | ||
| BI759672.1 | 4 | 823 | ||
| FJ224314.1 | 3 | 631 | ||
| KM220603.1 | ||||
| KM220604.1 | 3 | 620 | ||
| KM220605.1 | ||||
| KM220606.1 | 3 | 630 | ||
| M24906.1 | 1 | 288 | AAA60267.1 | |
| S45630.1 | 3 | 631 | AAB23453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482162 | Chr.11:111908626 - 111911749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map