Assay Details
Target Gene Details
Entrez Gene ID: | 8544 |
Gene Name: | pirin |
Gene Aliases: |
- |
Location: |
Chr.X:15384799-15493588 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIR | NM_001018109.2 | NP_001018119.1 | ||
| NM_003662.3 | NP_003653.1 | |||
| AK309656.1 | ||||
| BC002517.2 | AAH02517.1 | |||
| BT019583.1 | AAV38390.1 | |||
| BT019584.1 | AAV38391.1 | |||
| CR541822.1 | CAG46621.1 | |||
| Y07867.1 | CAA69194.1 | |||
| Y07868.1 | CAA69195.1 |
Target Gene Details
Entrez Gene ID: | 100532742 |
Gene Name: | PIR-FIGF readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:15345591-15491309 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIR-FIGF | NR_037859.2 | |||
| AX740234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515969 | Chr.X:15388939 - 15406564 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map