Assay Details
Target Gene Details
Entrez Gene ID: | 79645 |
Gene Name: | EF-hand calcium binding domain 1 |
Gene Aliases: |
- |
Location: |
Chr.8:48714915-48735843 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB1 | NM_001142857.1 | NP_001136329.1 | ||
| NM_024593.3 | 6 | 1352 | NP_078869.1 | |
| NR_024605.1 | 6 | 1068 | ||
| XM_005251303.1 | 7 | 1228 | XP_005251360.1 | |
| XM_011517589.1 | XP_011515891.1 | |||
| XM_011517590.2 | 7 | 1482 | XP_011515892.1 | |
| XM_011517591.2 | 7 | 1455 | XP_011515893.1 | |
| XM_017013826.1 | 6 | 1129 | XP_016869315.1 | |
| XM_017013827.1 | 8 | 1605 | XP_016869316.1 | |
| AK021829.1 | 6 | 1210 | BAB13906.1 | |
| AK299659.1 | ||||
| AK307101.1 | ||||
| AK316037.1 | 5 | 1148 | ||
| BC025676.1 | 6 | 1273 | AAH25676.1 |
More Information
Set Membership: |
|

Genomic Map