Assay Details
Target Gene Details
Entrez Gene ID: | 7158 |
Gene Name: | tumor protein p53 binding protein 1 |
Gene Aliases: |
53BP1, TDRD30, TP53, p202, p53BP1 |
Location: |
Chr.15:43403064-43510509 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP1 | XM_011521985.1 | 27 | 7754 | XP_011520287.1 |
| XM_017022533.1 | 29 | 8009 | XP_016878022.1 | |
| AK125173.1 | 1 | 768 |
Target Gene Details
Entrez Gene ID: | 27229 |
Gene Name: | tubulin gamma complex associated protein 4 |
Gene Aliases: |
76P, GCP-4, GCP4, Grip76, MCCRP3 |
Location: |
Chr.15:43371059-43409771 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBGCP4 | NM_001286414.2 | 18 | 2611 | NP_001273343.1 |
| NM_014444.4 | 18 | 2608 | NP_055259.2 | |
| XM_011521454.2 | 17 | 2494 | XP_011519756.1 | |
| XM_011521455.2 | 18 | 2509 | XP_011519757.1 | |
| XM_017022078.1 | 17 | 2491 | XP_016877567.1 | |
| AK001639.1 | 14 | 1922 | BAA91802.1 | |
| AK026551.1 | 18 | 2598 | ||
| AK027703.1 | 18 | 2569 | ||
| BC012801.2 | 18 | 2552 | AAH12801.1 |
More Information
Set Membership: |
|

Genomic Map