Assay Details
Target Gene Details
Entrez Gene ID: | 51114 |
Gene Name: | zinc finger DHHC-type containing 9 |
Gene Aliases: |
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380 |
Location: |
Chr.X:129803288-129843934 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC9 | NM_001008222.2 | 10 | 1607 | NP_001008223.1 |
| NM_016032.3 | 11 | 1587 | NP_057116.2 | |
| AB209807.1 | 10 | 1526 | BAD93044.1 | |
| AF151847.1 | 11 | 1475 | AAD34084.1 | |
| AK001112.1 | 11 | 1537 | ||
| AK001424.1 | 8 | 921 | BAA91683.1 | |
| AK001524.1 | 10 | 1338 | BAA91740.1 | |
| AK096967.1 | 11 | 1238 | ||
| AL161962.1 | 11 | 1464 | CAB82308.1 | |
| AY358558.1 | 11 | 1519 | AAQ88922.1 | |
| AY952881.8 | 11 | 1559 | ||
| BC000035.2 | 10 | 1481 | AAH00035.1 | |
| BC003128.1 | 11 | 1525 | AAH03128.1 | |
| BC006200.2 | 11 | 1521 | AAH06200.1 | |
| BC012826.2 | 11 | 1521 | AAH12826.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
| nsv526197 | Chr.X:129672435 - 129826254 on Build GRCh38 | Gain |
|
| esv2758893 | Chr.X:129563598 - 129830109 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map