Assay Details
Target Gene Details
Entrez Gene ID: | 3437 |
Gene Name: | interferon induced protein with tetratricopeptide repeats 3 |
Gene Aliases: |
CIG-49, GARG-49, IFI60, IFIT4, IRG2, ISG60, P60, RIG-G, cig41 |
Location: |
Chr.10:89327819-89340968 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFIT3 | NM_001031683.3 | 2 | 1150 | NP_001026853.1 |
| NM_001289758.1 | 3 | 1379 | NP_001276687.1 | |
| NM_001289759.1 | 3 | 1272 | NP_001276688.1 | |
| NM_001549.5 | 2 | 1261 | NP_001540.2 | |
| AF026939.1 | 2 | 1100 | AAB95160.1 | |
| AF083470.1 | 2 | 1084 | AAC63524.1 | |
| AK290427.1 | 2 | 1171 | ||
| AK297137.1 | 3 | 1070 | ||
| AK312675.1 | 2 | 1081 | ||
| BC001383.2 | 2 | 1083 | AAH01383.1 | |
| BC004977.1 | 2 | 1056 | AAH04977.1 | |
| BT007284.1 | 1 | 1006 | AAP35948.1 | |
| U52513.1 | 2 | 1067 | AAB40606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34085 | Chr.10:89306192 - 89414418 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map