Assay Details
Target Gene Details
Entrez Gene ID: | 64210 |
Gene Name: | MMS19 homolog, cytosolic iron-sulfur assembly component |
Gene Aliases: |
MET18, MMS19L, hMMS19 |
Location: |
Chr.10:97458324-97498794 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
More Information
Set Membership: |
|

Genomic Map