Assay Details
Target Gene Details
Entrez Gene ID: | 332 |
Gene Name: | baculoviral IAP repeat containing 5 |
Gene Aliases: |
API4, EPR-1 |
Location: |
Chr.17:78214196-78225635 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIRC5 | NM_001012270.1 | 3 | 2322 | NP_001012270.1 |
| NM_001012271.1 | 5 | 2509 | NP_001012271.1 | |
| NM_001168.2 | 4 | 2440 | NP_001159.2 | |
| AI800528.1 | 1 | 216 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524306 | Chr.17:78183449 - 78280693 on Build GRCh38 | Gain |
|
| nsv952373 | Chr.17:78214820 - 78283319 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map