Assay Details
Target Gene Details
Entrez Gene ID: | 5873 |
Gene Name: | RAB27A, member RAS oncogene family |
Gene Aliases: |
GS2, HsT18676, RAB27, RAM |
Location: |
Chr.15:55202966-55291188 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB27A | NM_004580.4 | NP_004571.2 | ||
| NM_183234.2 | NP_899057.1 | |||
| NM_183235.2 | NP_899058.1 | |||
| NM_183236.2 | NP_899059.1 | |||
| XM_005254576.4 | XP_005254633.1 | |||
| XM_011521852.1 | XP_011520154.1 | |||
| XM_011521854.1 | XP_011520156.1 | |||
| XM_011521855.2 | XP_011520157.1 | |||
| XM_011521856.2 | XP_011520158.1 | |||
| AF125393.1 | AAD43049.1 | |||
| AF498953.1 | AAM21101.1 | |||
| AK312836.1 | ||||
| BC107680.1 | AAI07681.1 | |||
| BC132800.1 | ||||
| BC136423.1 | ||||
| BM014705.1 | ||||
| CR457076.1 | CAG33357.1 | |||
| CR536496.1 | CAG38735.1 | |||
| CR541693.1 | CAG46494.1 | |||
| U38654.3 | AAC50271.2 | |||
| U57094.1 | AAC51195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761899 | Chr.15:53592144 - 55643724 on Build GRCh38 | Gain+Loss |
|
| nsv1039471 | Chr.15:54845408 - 55260357 on Build GRCh38 | Gain |
|
| nsv1043789 | Chr.15:55108674 - 55800256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map