Assay Details
Target Gene Details
Entrez Gene ID: | 10536 |
Gene Name: | prolyl 3-hydroxylase 3 |
Gene Aliases: |
GRCB, HSU47926, LEPREL2 |
Location: |
Chr.12:6828373-6839854 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3H3 | NM_014262.4 | 15 | 2237 | NP_055077.2 |
| AB209320.1 | 13 | 2004 | BAD92557.1 | |
| AJ430349.1 | 16 | 2466 | CAD23037.1 | |
| AK025976.1 | 15 | 2204 | ||
| AK096771.1 | 1 | 1761 | ||
| AK126766.1 | 16 | 2799 | ||
| AK297917.1 | 5 | 747 | ||
| BC017217.2 | 15 | 1778 | AAH17217.2 | |
| BC067251.1 | 15 | 1935 | AAH67251.1 | |
| BC080630.1 | 15 | 2010 | AAH80630.1 | |
| BQ009002.1 | 1 | 454 | ||
| U47926.1 | 14 | 1702 | AAC50464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526312 | Chr.12:6839304 - 6839735 on Build GRCh38 | Loss |
|
| nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss |
|
| esv3580405 | Chr.12:6826466 - 6903969 on Build GRCh38 | Gain |
|
| nsv516726 | Chr.12:6838864 - 6839718 on Build GRCh38 | Loss |
|
| nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map