Assay Details
Target Gene Details
Entrez Gene ID: | 66008 |
Gene Name: | trafficking kinesin protein 2 |
Gene Aliases: |
ALS2CR3, CALS-C, GRIF-1, GRIF1, MILT2, OIP98 |
Location: |
Chr.2:201377207-201451596 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAK2 | NM_015049.2 | 16 | 6095 | NP_055864.2 |
| XM_011511690.1 | 16 | 5939 | XP_011509992.1 | |
| XM_017004772.1 | 15 | 5999 | XP_016860261.1 | |
| AB011121.1 | 5 | 4314 | BAA25475.1 | |
| AB038951.1 | 16 | 6030 | BAB32501.1 |
More Information
Set Membership: |
|

Genomic Map