Assay Details
Target Gene Details
Entrez Gene ID: | 5778 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 7 |
Gene Aliases: |
BPTP-4, HEPTP, LC-PTP, LPTP, PTPNI |
Location: |
Chr.1:202147012-202161588 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN7 | NM_001199797.1 | 10 | 2393 | NP_001186726.1 |
| NM_002832.3 | 10 | 3239 | NP_002823.3 | |
| NM_080588.2 | 9 | 2732 | NP_542155.1 | |
| NR_037663.1 | 10 | 3186 | ||
| NR_037664.1 | 9 | 3055 | ||
| XM_011509819.1 | 11 | 2712 | XP_011508121.1 | |
| XM_011509820.2 | 11 | 2416 | XP_011508122.1 | |
| XM_011509821.1 | 11 | 2344 | XP_011508123.1 | |
| XM_011509822.2 | 11 | 2215 | XP_011508124.1 | |
| XM_011509824.1 | 10 | 2220 | XP_011508126.1 | |
| XM_017001935.1 | 11 | 2709 | XP_016857424.1 | |
| XM_017001936.1 | 10 | 2571 | XP_016857425.1 | |
| XM_017001937.1 | 10 | 2389 | XP_016857426.1 | |
| AK127214.1 | 10 | 3232 | ||
| BC001746.1 | 9 | 2732 | AAH01746.2 | |
| D11327.1 | 11 | 2160 | BAA01946.1 | |
| M64322.1 | 10 | 2235 | AAA59531.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115477816] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map