Assay Details
Target Gene Details
Entrez Gene ID: | 130120 |
Gene Name: | regenerating family member 3 gamma |
Gene Aliases: |
LPPM429, PAP IB, PAP-1B, PAP1B, PAPIB, REG III, REG-III, UNQ429 |
Location: |
Chr.2:79025686-79028504 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REG3G | NM_001008387.2 | 6 | 787 | NP_001008388.1 |
| NM_001270040.1 | 5 | 649 | NP_001256969.1 | |
| NM_198448.3 | 6 | 687 | NP_940850.1 | |
| XM_005264135.2 | 6 | 849 | XP_005264192.1 | |
| AB161037.1 | 6 | 672 | BAD51394.1 | |
| AB161038.1 | 6 | 772 | BAD51396.1 | |
| AI025191.1 | 1 | 175 | ||
| AK292595.1 | 6 | 671 | ||
| AY359047.1 | 6 | 673 | AAQ89406.1 | |
| AY428734.1 | 6 | 646 | AAR88147.1 | |
| BC103852.2 | AAI03853.1 | |||
| BC103853.1 | AAI03854.1 | |||
| BC103854.1 | AAI03855.1 | |||
| BI792387.1 | 1 | 164 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584761 | Chr.2:78317894 - 79203502 on Build GRCh38 | Loss |
|
| esv3892770 | Chr.2:78806791 - 79322248 on Build GRCh38 | Gain |
|
| nsv522260 | Chr.2:78312540 - 79523465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map