Assay Details
Target Gene Details
Entrez Gene ID: | 54434 |
Gene Name: | slingshot protein phosphatase 1 |
Gene Aliases: |
SSH1L |
Location: |
Chr.12:108782690-108857606 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSH1 | NM_018984.3 | 15 | 3256 | NP_061857.3 |
| XM_005268984.4 | 15 | 3967 | XP_005269041.1 | |
| XM_005268985.2 | 14 | 3082 | XP_005269042.1 | |
| XM_011538497.1 | 14 | 3348 | XP_011536799.1 | |
| XM_011538499.1 | 13 | 3147 | XP_011536801.1 | |
| XM_011538500.2 | 14 | 3172 | XP_011536802.1 | |
| XM_011538501.2 | 14 | 3168 | XP_011536803.1 | |
| XM_017019491.1 | 14 | 4194 | XP_016874980.1 | |
| XM_017019492.1 | 15 | 3309 | XP_016874981.1 | |
| XM_017019493.1 | 13 | 3053 | XP_016874982.1 | |
| XM_017019494.1 | 13 | 3148 | XP_016874983.1 | |
| AB037719.1 | 5 | 2291 | BAA92536.1 | |
| AB072355.1 | 15 | 3169 | BAB84114.1 | |
| BC062341.1 | 15 | 3248 | AAH62341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763002 | Chr.12:108780333 - 108811466 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map