Assay Details
Target Gene Details
Entrez Gene ID: | 51406 |
Gene Name: | nucleolar protein 7 |
Gene Aliases: |
C6orf90, PQBP3, RARG-1, dJ223E5.2 |
Location: |
Chr.6:13615327-13632470 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL7 | NM_001317724.1 | 6 | 603 | NP_001304653.1 |
| NM_016167.4 | 6 | 603 | NP_057251.2 | |
| AF172066.3 | 6 | 603 | AAD49745.2 | |
| AK075301.1 | 6 | 586 | BAC11534.1 | |
| AL050299.1 | 5 | 303 | CAB43398.1 | |
| BC023517.2 | 6 | 578 | AAH23517.1 | |
| BC062683.1 | 6 | 599 | AAH62683.1 | |
| BU071935.1 | 6 | 356 |
More Information
Set Membership: |
|

Genomic Map