Assay Details
Target Gene Details
Entrez Gene ID: | 80351 |
Gene Name: | tankyrase 2 |
Gene Aliases: |
ARTD6, PARP-5b, PARP-5c, PARP5B, PARP5C, TANK2, TNKL, pART6 |
Location: |
Chr.10:91798394-91865475 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNKS2 | NM_025235.3 | 8 | 1213 | NP_079511.1 |
| XM_005270185.4 | 8 | 1244 | XP_005270242.2 | |
| XM_011540213.1 | 8 | 1244 | XP_011538515.1 | |
| XM_017016696.1 | 8 | 1181 | XP_016872185.1 | |
| XM_017016697.1 | 8 | 1197 | XP_016872186.1 | |
| XM_017016698.1 | 8 | 908 | XP_016872187.1 | |
| XM_017016699.1 | 8 | 1197 | XP_016872188.1 | |
| XM_017016700.1 | 8 | 794 | XP_016872189.1 | |
| XM_017016701.1 | 8 | 1244 | XP_016872190.1 | |
| AF264912.1 | 8 | 916 | AAG44694.1 | |
| AF305081.1 | 8 | 1218 | AAG25674.1 | |
| AF309033.1 | 8 | 1167 | AAK82330.1 | |
| AF329696.1 | 8 | 1055 | AAK13463.1 | |
| AF342982.1 | 8 | 916 | AAK25811.1 | |
| AF438201.1 | 8 | 1295 | AAL40795.1 | |
| AK314612.1 | 8 | 1181 |
More Information
Set Membership: |
|

Genomic Map