Assay Details
Target Gene Details
Entrez Gene ID: | 4821 |
Gene Name: | NK2 homeobox 2 |
Gene Aliases: |
NKX2.2, NKX2B |
Location: |
Chr.20:21511010-21514026 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-2 | NM_002509.3 | 2 | 1423 | NP_002500.1 |
| XM_006723566.3 | 2 | 1080 | XP_006723629.1 | |
| CA772329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2659363 | Chr.20:21505907 - 21515954 on Build GRCh38 | Deletion |
|
| nsv952998 | Chr.20:21504663 - 21524162 on Build GRCh38 | Deletion |
|
| nsv833941 | Chr.20:21393666 - 21576223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map