Assay Details
Target Gene Details
Entrez Gene ID: | 23408 |
Gene Name: | sirtuin 5 |
Gene Aliases: |
SIR2L5 |
Location: |
Chr.6:13574222-13615158 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRT5 | NM_001193267.2 | NP_001180196.1 | ||
| NM_001242827.1 | NP_001229756.1 | |||
| NM_012241.4 | NP_036373.1 | |||
| NM_031244.3 | NP_112534.1 | |||
| XM_005248968.4 | XP_005249025.1 | |||
| XM_011514444.2 | XP_011512746.1 | |||
| XM_017010621.1 | XP_016866110.1 | |||
| XM_017010622.1 | XP_016866111.1 | |||
| XM_017010623.1 | XP_016866112.1 | |||
| AF083110.1 | AAD40853.1 | |||
| AJ420496.1 | ||||
| AK000355.1 | BAA91107.1 | |||
| AK294162.1 | ||||
| AK302467.1 | ||||
| BC000126.1 | AAH00126.1 | |||
| BX538030.1 | CAD97975.1 | |||
| CA406695.1 | ||||
| CB155518.1 | ||||
| DC314190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608129 | Chr.6:13525301 - 13607816 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map