Assay Details
Target Gene Details
Entrez Gene ID: | 143884 |
Gene Name: | CWF19-like 2, cell cycle control (S. pombe) |
Gene Aliases: |
- |
Location: |
Chr.11:107326345-107457846 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWF19L2 | NM_152434.2 | 18 | 2869 | NP_689647.2 |
| XM_017017239.1 | 17 | 2187 | XP_016872728.1 | |
| AI148333.1 | 1 | 417 | ||
| AK056905.1 | 16 | 2521 | BAB71307.1 | |
| AK126881.1 | 18 | 2860 | ||
| AK293001.1 | ||||
| BC028043.2 | 5 | 718 | AAH28043.1 | |
| BC056241.1 | 6 | 964 | AAH56241.1 | |
| BX647832.1 | ||||
| BX648704.1 | 18 | 2848 | ||
| BX649022.1 | 18 | 2839 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627644 | Chr.11:107257370 - 107361314 on Build GRCh38 | Gain |
|
| nsv556280 | Chr.11:107288014 - 107360879 on Build GRCh38 | Gain |
|
| nsv1035911 | Chr.11:107150265 - 107327455 on Build GRCh38 | Gain |
|
| nsv832261 | Chr.11:107275175 - 107450836 on Build GRCh38 | Loss |
|
| esv3580083 | Chr.11:107148818 - 107328113 on Build GRCh38 | Gain |
|
| esv3627642 | Chr.11:107070173 - 107331876 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map