Assay Details
Target Gene Details
Entrez Gene ID: | 55088 |
Gene Name: | coiled-coil domain containing 186 |
Gene Aliases: |
C10orf118 |
Location: |
Chr.10:114120862-114174312 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC186 | NM_001321829.1 | 12 | 2155 | NP_001308758.1 |
| NM_018017.3 | 11 | 2030 | NP_060487.2 | |
| NR_135815.1 | 12 | 2155 | ||
| XM_011539915.2 | 10 | 1373 | XP_011538217.1 | |
| AF273054.1 | 10 | 1545 | AAM44457.1 | |
| AK057508.1 | 6 | 648 | ||
| AK307517.1 | 11 | 2007 | ||
| AL834368.1 | 10 | 1229 | CAD39031.2 | |
| BC022229.1 | 1 | 54 | ||
| BC100670.1 | 12 | 2082 | AAI00671.1 | |
| BC103499.1 | 11 | 1957 | AAI03500.1 | |
| BC105632.1 | 11 | 1957 | AAI05633.1 | |
| BC152306.1 | 11 | 1961 | ||
| BX537964.1 | 11 | 2002 | CAD97928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052267 | Chr.10:114103969 - 114155081 on Build GRCh38 | Loss |
|
| nsv1045020 | Chr.10:114131051 - 114164562 on Build GRCh38 | Loss |
|

Genomic Map