Assay Details
Target Gene Details
Entrez Gene ID: | 51305 |
Gene Name: | potassium two pore domain channel subfamily K member 9 |
Gene Aliases: |
K2p9.1, KT3.2, TASK-3, TASK3 |
Location: |
Chr.8:139600838-139703135 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK9 | NM_001282534.1 | 2 | 1142 | NP_001269463.1 |
| NR_104210.1 | 2 | 1142 | ||
| XM_011517101.2 | XP_011515403.1 | |||
| XM_011517102.2 | 2 | 1221 | XP_011515404.1 | |
| XM_011517103.2 | XP_011515405.1 | |||
| XM_017013530.1 | 2 | 937 | XP_016869019.1 | |
| XM_017013531.1 | 2 | 908 | XP_016869020.1 | |
| AF212829.1 | 2 | 1078 | AAF63708.1 | |
| AF248241.1 | 2 | 1142 | AAG31730.1 | |
| AF257080.1 | 2 | 1084 | AAG33126.1 | |
| AF279809.1 | 2 | 1078 | AAF85982.1 | |
| AY190605.1 | 2 | 1078 | AAO38739.1 | |
| BC075079.2 | 2 | 1127 | AAH75079.1 | |
| BC075080.2 | 2 | 1127 | AAH75080.1 | |
| BC112063.1 | 2 | 1127 | AAI12064.1 | |
| BC112065.1 | 2 | 1127 | AAI12066.1 |
More Information
Set Membership: |
|

Genomic Map