Assay Details
Target Gene Details
Entrez Gene ID: | 146050 |
Gene Name: | zinc finger and SCAN domain containing 29 |
Gene Aliases: |
ZNF690, Zfp690 |
Location: |
Chr.15:43358172-43371032 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN29 | NM_152455.3 | 5 | 4514 | NP_689668.3 |
| XM_006720401.3 | 6 | 5274 | XP_006720464.1 | |
| XM_011521266.2 | 6 | 4954 | XP_011519568.1 | |
| AK023525.1 | 1 | 774 | ||
| AK098289.1 | 1 | 2119 | ||
| AK123753.1 | 1 | 1455 | ||
| BC017179.2 | 3 | 3380 | AAH17179.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1512 | Chr.15:43324636 - 43369448 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map