Assay Details
Target Gene Details
Entrez Gene ID: | 2999 |
Gene Name: | granzyme H |
Gene Aliases: |
CCP-X, CGL-2, CSP-C, CTLA1, CTSGL2 |
Location: |
Chr.14:24606480-24609763 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GZMH | NM_001270780.1 | NP_001257709.1 | ||
| NM_001270781.1 | NP_001257710.1 | |||
| NM_033423.4 | NP_219491.1 | |||
| XM_011536683.2 | XP_011534985.1 | |||
| AY232657.1 | AAP70247.1 | |||
| AY232658.1 | AAP70248.1 | |||
| BC027974.1 | AAH27974.1 | |||
| BQ054303.1 | ||||
| CD000418.1 | ||||
| M36118.1 | AAA03248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748495 | Chr.14:24607596 - 24632432 on Build GRCh38 | Deletion |
|
| nsv510631 | Chr.14:24575977 - 24703105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map