Assay Details
Target Gene Details
Entrez Gene ID: | 64175 |
Gene Name: | prolyl 3-hydroxylase 1 |
Gene Aliases: |
GROS1, LEPRE1, OI8 |
Location: |
Chr.1:42746335-42767084 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3H1 | NM_001146289.1 | NP_001139761.1 | ||
| NM_001243246.1 | NP_001230175.1 | |||
| NM_022356.3 | NP_071751.3 | |||
| XM_005271110.2 | XP_005271167.1 | |||
| XM_011541948.1 | XP_011540250.1 | |||
| XM_017002051.1 | XP_016857540.1 | |||
| XM_017002052.1 | XP_016857541.1 | |||
| AF097431.1 | AAG31018.1 | |||
| AF097432.1 | AAG31019.1 | |||
| AK025841.1 | BAB15256.1 | |||
| AK027648.1 | BAB55264.1 | |||
| AK027680.1 | BAB55291.1 | |||
| AK027697.1 | BAB55305.1 | |||
| AK075418.1 | BAC11608.1 | |||
| AK297980.1 | ||||
| AK300210.1 | ||||
| BC015309.2 | AAH15309.2 | |||
| BC108311.1 | AAI08312.1 | |||
| BT007039.1 | AAP35688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950700 | Chr.1:42731330 - 42756229 on Build GRCh38 | Deletion |
|
| esv3577249 | Chr.1:42576524 - 42885152 on Build GRCh38 | Gain |
|
| nsv829682 | Chr.1:42589429 - 42773568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map