Assay Details
Target Gene Details
Entrez Gene ID: | 10602 |
Gene Name: | CDC42 effector protein 3 |
Gene Aliases: |
BORG2, CEP3, UB1 |
Location: |
Chr.2:37641882-37672535 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP3 | NM_001270436.1 | 2 | 242 | NP_001257365.1 |
| NM_001270437.1 | 2 | 198 | NP_001257366.1 | |
| NM_001270438.1 | 2 | 174 | NP_001257367.1 | |
| NM_006449.4 | 2 | 833 | NP_006440.2 | |
| XM_011532482.2 | 2 | 639 | XP_011530784.1 | |
| AK128735.1 | 2 | 260 | ||
| AK313483.1 | 2 | 277 | ||
| AL136842.1 | 2 | 792 | CAB66776.1 | |
| AL538577.3 | 2 | 826 | ||
| BC019270.1 | 2 | 261 | AAH19270.1 | |
| BQ067953.1 | 2 | 174 | ||
| DB121680.1 | 2 | 167 | ||
| HY003915.1 | 2 | 174 | ||
| HY017509.1 | 2 | 245 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833825 | Chr.2:37581026 - 37758774 on Build GRCh38 | Gain |
|
| esv2759043 | Chr.2:37569511 - 37911796 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map