Assay Details
Target Gene Details
Entrez Gene ID: | 26003 |
Gene Name: | golgi reassembly stacking protein 2 |
Gene Aliases: |
GOLPH6, GRASP55, GRS2, p59 |
Location: |
Chr.2:170928438-170967135 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GORASP2 | NM_001201428.1 | NP_001188357.1 | ||
| NM_015530.4 | NP_056345.3 | |||
| XM_006712408.2 | XP_006712471.1 | |||
| AK023082.1 | BAB14395.1 | |||
| AK023201.1 | BAB14459.1 | |||
| AK027349.1 | BAB55054.1 | |||
| AK125761.1 | ||||
| AK225810.1 | ||||
| AK293640.1 | ||||
| AK296698.1 | ||||
| AK298020.1 | ||||
| AK298771.1 | ||||
| AK315508.1 | ||||
| AL117430.1 | CAB55919.1 | |||
| BC007770.2 | AAH07770.1 | |||
| DC399516.1 |
More Information
Set Membership: |
|

Genomic Map