Assay Details
Target Gene Details
Entrez Gene ID: | 5469 |
Gene Name: | mediator complex subunit 1 |
Gene Aliases: |
CRSP1, CRSP200, DRIP205, DRIP230, PBP, PPARBP, PPARGBP, RB18A, TRAP220, TRIP2 |
Location: |
Chr.17:39404285-39451281 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED1 | NM_004774.3 | 17 | 4662 | NP_004765.2 |
| XM_005257465.3 | 16 | 4536 | XP_005257522.1 | |
| XM_006721957.1 | XP_006722020.1 | |||
| XM_017024779.1 | 16 | 4595 | XP_016880268.1 | |
| AF055994.1 | 17 | 4642 | AAC39854.1 | |
| AF283812.1 | 16 | 4405 | AAF98352.1 | |
| AK299789.1 | 16 | 4527 | ||
| AK314185.1 | 17 | 4659 | ||
| BC060758.1 | AAH60758.1 | |||
| BC131783.1 | 17 | 4516 | ||
| Y13467.1 | 17 | 4640 | CAA73867.1 |
More Information
Set Membership: |
|

Genomic Map