Assay Details
Target Gene Details
Entrez Gene ID: | 4839 |
Gene Name: | NOP2 nucleolar protein |
Gene Aliases: |
NOL1, NOP120, NSUN1, p120 |
Location: |
Chr.12:6556870-6568332 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP2 | NM_001033714.2 | 11 | 1319 | NP_001028886.1 |
| NM_001258308.1 | 11 | 1263 | NP_001245237.1 | |
| NM_001258309.1 | 12 | 1362 | NP_001245238.1 | |
| NM_001258310.1 | 11 | 1251 | NP_001245239.1 | |
| NM_006170.3 | 11 | 1251 | NP_006161.2 | |
| XM_005253691.1 | 11 | 1296 | XP_005253748.1 | |
| AK056208.1 | 11 | 1233 | ||
| AK129852.1 | 11 | 1194 | ||
| AK307582.1 | 11 | 1245 | ||
| BC000656.2 | 11 | 1184 | AAH00656.1 | |
| BC065257.1 | 11 | 1261 | ||
| BC082985.1 | 11 | 1201 | ||
| BC106072.1 | 12 | 1308 | AAI06073.1 | |
| BC128183.1 | 11 | 1238 | ||
| BC128184.1 | 11 | 1238 | ||
| M32110.1 | 10 | 1177 | AAA36398.1 | |
| X55504.1 | 10 | 1133 | CAA39119.1 |
More Information
Set Membership: |
|

Genomic Map