Assay Details
Target Gene Details
Entrez Gene ID: | 7805 |
Gene Name: | lysosomal protein transmembrane 5 |
Gene Aliases: |
CLAST6 |
Location: |
Chr.1:30732468-30757840 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAPTM5 | NM_006762.2 | NP_006753.1 | ||
| XM_011542098.2 | XP_011540400.1 | |||
| AK292009.1 | ||||
| AK297014.1 | ||||
| BC106896.1 | AAI06897.1 | |||
| BC106897.1 | AAI06898.1 | |||
| CT003824.1 | ||||
| D42042.1 | ||||
| U30498.1 | AAA74018.1 | |||
| U51240.1 | AAB08975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv145n100 | Chr.1:30666071 - 30792998 on Build GRCh38 | Gain |
|
| nsv509124 | Chr.1:30710610 - 30759706 on Build GRCh38 | Insertion |
|
| nsv545947 | Chr.1:30640876 - 30799204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map