Assay Details
Target Gene Details
Entrez Gene ID: | 11153 |
Gene Name: | FIC domain containing |
Gene Aliases: |
HIP13, HYPE, UNQ3041 |
Location: |
Chr.12:108515274-108519603 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FICD | NM_007076.2 | 3 | 532 | NP_009007.2 |
| AK000014.1 | 3 | 506 | ||
| AK026074.1 | 3 | 532 | ||
| AY358992.1 | 3 | 501 | AAQ89351.1 | |
| BC001342.1 | 3 | 490 | AAH01342.2 |
More Information
Set Membership: |
|

Genomic Map