Assay Details
Target Gene Details
Entrez Gene ID: | 200186 |
Gene Name: | CREB regulated transcription coactivator 2 |
Gene Aliases: |
TORC-2, TORC2 |
Location: |
Chr.1:153947672-153958656 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRTC2 | NM_181715.2 | 14 | 2370 | NP_859066.1 |
| XM_005244946.1 | 14 | 2366 | XP_005245003.1 | |
| XM_005244949.2 | 14 | 2243 | XP_005245006.1 | |
| XM_017000575.1 | 13 | 2139 | XP_016856064.1 | |
| XM_017000576.1 | 13 | 2124 | XP_016856065.1 | |
| XM_017000577.1 | 13 | 2268 | XP_016856066.1 | |
| XM_017000578.1 | 13 | 2253 | XP_016856067.1 | |
| XM_017000579.1 | 8 | 1665 | XP_016856068.1 | |
| AF289574.1 | 5 | 1329 | AAL55758.1 | |
| AK096382.1 | 12 | 1961 | ||
| AK223559.1 | BAD97279.1 | |||
| AY360172.1 | AAQ98857.1 | |||
| AY726597.1 | 14 | 2150 | ||
| BC028886.1 | 3 | 605 | AAH28886.1 | |
| BC053562.1 | 14 | 2281 | AAH53562.1 | |
| BM987537.1 | 1 | 327 |
More Information
Set Membership: |
|

Genomic Map