Assay Details
Target Gene Details
Entrez Gene ID: | 712 |
Gene Name: | complement C1q A chain |
Gene Aliases: |
- |
Location: |
Chr.1:22636625-22639682 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QA | NM_015991.2 | 2 | 137 | NP_057075.1 |
| AF135157.1 | 1 | 64 | AAD32626.1 | |
| AF260332.1 | 1 | 70 | AAG44663.1 | |
| AK311980.1 | 2 | 80 | ||
| BC030153.2 | 2 | 107 | AAH30153.1 | |
| BC071986.1 | 1 | 73 | AAH71986.1 | |
| BG548439.1 | 2 | 138 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34161 | Chr.1:22599762 - 22817730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map