Assay Details
Target Gene Details
Entrez Gene ID: | 102724058 |
Gene Name: | uncharacterized LOC102724058 |
Gene Aliases: |
- |
Location: |
Chr.2:165957418-166036400 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724058 | NR_110598.1 | |||
| HY008183.1 | ||||
| HY011842.1 | ||||
| HY029566.1 |
Target Gene Details
Entrez Gene ID: | 6323 |
Gene Name: | sodium voltage-gated channel alpha subunit 1 |
Gene Aliases: |
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI |
Location: |
Chr.2:165989160-166149216 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN1A | NM_001165963.1 | 26 | 5986 | NP_001159435.1 |
| NM_001165964.1 | 26 | 5902 | NP_001159436.1 | |
| NM_001202435.1 | 28 | 6195 | NP_001189364.1 | |
| NM_006920.4 | 26 | 5953 | NP_008851.3 | |
| XM_011511602.2 | 27 | 6262 | XP_011509904.1 | |
| XM_011511604.2 | 27 | 6122 | XP_011509906.1 | |
| XM_011511605.2 | 28 | 6326 | XP_011509907.1 | |
| XM_011511606.2 | 28 | 6270 | XP_011509908.1 | |
| XM_017004644.1 | 29 | 6441 | XP_016860133.1 | |
| XM_017004645.1 | 28 | 6329 | XP_016860134.1 | |
| XM_017004646.1 | 27 | 6243 | XP_016860135.1 | |
| XM_017004647.1 | 29 | 6415 | XP_016860136.1 | |
| XM_017004648.1 | 28 | 6192 | XP_016860137.1 | |
| XM_017004649.1 | 27 | 6154 | XP_016860138.1 | |
| XM_017004650.1 | 27 | 6228 | XP_016860139.1 | |
| XM_017004651.1 | 28 | 6133 | XP_016860140.1 | |
| XM_017004652.1 | 27 | 6173 | XP_016860141.1 | |
| XM_017004653.1 | 28 | 6280 | XP_016860142.1 | |
| XM_017004654.1 | 28 | 6361 | XP_016860143.1 | |
| AB093548.1 | 26 | 5968 | BAC21101.1 | |
| AB093549.1 | 26 | 5935 | BAC21102.1 | |
| AB098335.1 | 26 | 5884 | BAC45228.1 | |
| AF225985.1 | 26 | 5956 | AAK00217.1 | |
| AK293759.1 | 2 | 1244 | ||
| AK294900.1 | 6 | 1729 | ||
| AY043484.1 | 26 | 5935 | AAK95360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv961904 | Chr.2:165991129 - 165992860 on Build GRCh38 | Duplication |
|
| nsv834445 | Chr.2:165800387 - 165995152 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map