Assay Details
Target Gene Details
Entrez Gene ID: | 9241 |
Gene Name: | noggin |
Gene Aliases: |
SYM1, SYNS1, SYNS1A |
Location: |
Chr.17:56593699-56595590 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOG | NM_005450.4 | 1 | 1391 | NP_005441.1 |
| BC034027.1 | 1 | 1176 | AAH34027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672462 | Chr.17:56592584 - 56598331 on Build GRCh38 | Deletion |
|
| nsv952343 | Chr.17:56592840 - 56598239 on Build GRCh38 | Deletion |
|
| nsv833494 | Chr.17:56466715 - 56645877 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map