Assay Details
Target Gene Details
Entrez Gene ID: | 3936 |
Gene Name: | lymphocyte cytosolic protein 1 |
Gene Aliases: |
CP64, HEL-S-37, L-PLASTIN, LC64P, LPL, PLS2 |
Location: |
Chr.13:46125923-46182324 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCP1 | NM_002298.4 | 12 | 1547 | NP_002289.2 |
| XM_005266374.1 | 12 | 1445 | XP_005266431.1 | |
| AB208984.1 | 12 | 1392 | BAD92221.1 | |
| AK097275.1 | 2 | 834 | ||
| AK129625.1 | ||||
| AK223305.1 | 12 | 1482 | BAD97025.1 | |
| AK312393.1 | 12 | 1413 | ||
| BC007673.1 | 12 | 1385 | AAH07673.1 | |
| BC010271.1 | 12 | 1449 | AAH10271.1 | |
| CR936795.1 | 12 | 1435 | ||
| DA820090.1 | 1 | 65 | ||
| EU794648.1 | 12 | 1385 | ||
| J02923.1 | 12 | 1383 | AAA63236.1 | |
| M22300.1 | 10 | 1138 | AAB02845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| nsv832598 | Chr.13:45974228 - 46149047 on Build GRCh38 | Gain |
|
| nsv1049556 | Chr.13:46128266 - 46186795 on Build GRCh38 | Gain |
|
| nsv455876 | Chr.13:46044155 - 46739284 on Build GRCh38 | Gain |
|
| nsv1051215 | Chr.13:46135254 - 46243231 on Build GRCh38 | Gain |
|
| esv3631977 | Chr.13:46120553 - 46174096 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map