Assay Details
Target Gene Details
Entrez Gene ID: | 6258 |
Gene Name: | retinoid X receptor gamma |
Gene Aliases: |
NR2B3, RXRC |
Location: |
Chr.1:165400922-165445355 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RXRG | NM_001256570.1 | NP_001243499.1 | ||
| NM_001256571.1 | NP_001243500.1 | |||
| NM_006917.4 | NP_008848.1 | |||
| AB307706.1 | ||||
| AB593015.1 | ||||
| AB593016.1 | ||||
| AK312922.1 | ||||
| BC012063.1 | AAH12063.1 | |||
| BM728015.1 | ||||
| CR456705.1 | CAG32986.1 | |||
| HQ692845.1 | ||||
| U38480.1 | AAA80681.1 |
More Information
Set Membership: |
|

Genomic Map