Assay Details
Target Gene Details
Entrez Gene ID: | 387640 |
Gene Name: | SKI/DACH domain containing 1 |
Gene Aliases: |
C10orf140, DLN-1 |
Location: |
Chr.10:21513475-21525682 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKIDA1 | NM_207371.3 | 4 | 4759 | NP_997254.3 |
| XM_005252450.3 | 5 | 3868 | XP_005252507.1 | |
| XM_011519480.2 | 5 | 3865 | XP_011517782.1 | |
| XM_017016225.1 | 5 | 4659 | XP_016871714.1 | |
| XM_017016226.1 | 5 | 4662 | XP_016871715.1 | |
| AB234866.1 | 2 | 2357 | BAE93016.1 | |
| AK127130.1 | 2 | 1794 | BAC86842.1 | |
| AK131456.1 | 5 | 4489 | BAD18601.1 | |
| AL834446.1 | 1 | 1471 | CAD39106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv550201 | Chr.10:21507152 - 21535798 on Build GRCh38 | Loss |
|
| nsv517469 | Chr.10:21494705 - 21533927 on Build GRCh38 | Loss |
|
| nsv825289 | Chr.10:21473577 - 21556946 on Build GRCh38 | Loss |
|
| dgv133n67 | Chr.10:21450598 - 21585405 on Build GRCh38 | Gain |
|
| nsv527055 | Chr.10:21475090 - 21533927 on Build GRCh38 | Loss |
|
| nsv550200 | Chr.10:21493913 - 21535798 on Build GRCh38 | Loss |
|
| dgv135n67 | Chr.10:21488928 - 21542017 on Build GRCh38 | Gain |
|
| dgv134n67 | Chr.10:21468855 - 21549961 on Build GRCh38 | Gain |
|

Genomic Map