Assay Details
Target Gene Details
Entrez Gene ID: | 392309 |
Gene Name: | olfactory receptor family 13 subfamily J member 1 |
Gene Aliases: |
OR9-2 |
Location: |
Chr.9:35869463-35870401 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR13J1 | NM_001004487.1 | 1 | 603 | NP_001004487.1 |
| AY726589.1 | 1 | 522 | ||
| BC136715.1 | 1 | 666 | ||
| BC136716.1 | 1 | 666 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831546 | Chr.9:35826697 - 36044567 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map