Assay Details
Target Gene Details
Entrez Gene ID: | 995 |
Gene Name: | cell division cycle 25C |
Gene Aliases: |
CDC25, PPP1R60 |
Location: |
Chr.5:138285265-138338355 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC25C | NM_001287582.1 | 14 | 1723 | NP_001274511.1 |
| NM_001287583.1 | 14 | 1840 | NP_001274512.1 | |
| NM_001318098.1 | 14 | 1844 | NP_001305027.1 | |
| NM_001790.4 | 14 | 1727 | NP_001781.2 | |
| NM_022809.3 | 11 | 1508 | NP_073720.1 | |
| XM_005272145.3 | 13 | 1738 | XP_005272202.1 | |
| XM_006714739.3 | 13 | 1438 | XP_006714802.1 | |
| XM_011543759.2 | 15 | 1511 | XP_011542061.1 | |
| XM_011543760.2 | 12 | 1675 | XP_011542062.1 | |
| XM_011543761.2 | 11 | 1621 | XP_011542063.1 | |
| XM_011543762.1 | 11 | 1421 | XP_011542064.1 | |
| XM_011543763.1 | 12 | 1136 | XP_011542065.1 | |
| XM_011543764.1 | 11 | 1189 | XP_011542066.1 | |
| XM_017010101.1 | 13 | 1616 | XP_016865590.1 | |
| AJ304504.1 | CAC19192.1 | |||
| AK097710.1 | 14 | 1840 | ||
| AK301828.1 | 15 | 1790 | ||
| BC019089.2 | 14 | 1617 | AAH19089.1 | |
| DB516402.1 | ||||
| M34065.1 | 14 | 1616 | AAA35666.1 |
More Information
Set Membership: |
|

Genomic Map