Assay Details
Target Gene Details
Entrez Gene ID: | 6915 |
Gene Name: | thromboxane A2 receptor |
Gene Aliases: |
BDPLT13, TXA2-R |
Location: |
Chr.19:3594506-3608749 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBXA2R | NM_001060.5 | 2 | 465 | NP_001051.1 |
| NM_201636.2 | 2 | 465 | NP_963998.2 | |
| XM_011528214.2 | 3 | 2499 | XP_011526516.1 | |
| AY429110.1 | 1 | 78 | AAR07905.1 | |
| BC028027.1 | 2 | 465 | ||
| BC074749.2 | 1 | 117 | AAH74749.1 | |
| BC074750.2 | 1 | 117 | AAH74750.1 | |
| BC121148.1 | 1 | 112 | ||
| U11271.1 | 2 | 290 | AAA58957.1 | |
| U27325.1 | 2 | 201 | AAA68608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv833713 | Chr.19:3564423 - 3627361 on Build GRCh38 | Loss |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map