Assay Details
Target Gene Details
Entrez Gene ID: | 5394 |
Gene Name: | exosome component 10 |
Gene Aliases: |
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4 |
Location: |
Chr.1:11066613-11099910 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC10 | NM_001001998.2 | 22 | 2487 | NP_001001998.1 |
| NM_002685.3 | 21 | 2412 | NP_002676.1 | |
| XM_005263475.2 | 22 | 2461 | XP_005263532.2 | |
| XM_017001494.1 | 21 | 2187 | XP_016856983.1 | |
| XM_017001495.1 | 21 | 2172 | XP_016856984.1 | |
| XM_017001496.1 | 22 | 2532 | XP_016856985.1 | |
| XM_017001497.1 | 22 | 2517 | XP_016856986.1 | |
| AB209236.1 | 18 | 4019 | BAD92473.1 | |
| AK093955.1 | 2 | 2219 | ||
| AK123365.1 | 10 | 1660 | ||
| AK296559.1 | 20 | 2987 | ||
| BC009908.2 | 18 | 1809 | AAH09908.2 | |
| BC039901.1 | 22 | 2428 | AAH39901.1 | |
| BC073788.1 | 22 | 2463 | AAH73788.1 | |
| CA307576.1 | 4 | 373 | ||
| L01457.1 | 21 | 2371 | AAB59352.1 | |
| X66113.1 | 22 | 2445 | CAA46904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map