Assay Details
Target Gene Details
Entrez Gene ID: | 390191 |
Gene Name: | olfactory receptor family 5 subfamily B member 12 |
Gene Aliases: |
OR11-241, OR5B12P, OR5B16, OST743 |
Location: |
Chr.11:58439120-58440173 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR5B12 | NM_001004733.2 | 1 | 700 | NP_001004733.1 |
| BC136947.1 | 1 | 700 | ||
| BC136948.1 | 1 | 700 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517849 | Chr.11:58361580 - 58496708 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map