Assay Details
Target Gene Details
Entrez Gene ID: | 57556 |
Gene Name: | semaphorin 6A |
Gene Aliases: |
HT018, SEMA, SEMA6A1, SEMAQ, VIA |
Location: |
Chr.5:116443555-116574926 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6A | NM_001300780.1 | 20 | 2939 | NP_001287709.1 |
| NM_020796.4 | 19 | 2888 | NP_065847.1 | |
| XM_005272042.3 | 18 | 2287 | XP_005272099.2 | |
| XM_006714663.1 | 20 | 2853 | XP_006714726.1 | |
| XM_017009675.1 | 20 | 2258 | XP_016865164.1 | |
| XM_017009676.1 | 15 | 4805 | XP_016865165.1 | |
| AB037789.1 | 20 | 2408 | ||
| AF279656.1 | 19 | 2765 | AAG29378.1 | |
| AK027439.1 | 6 | 665 | BAB55111.1 | |
| AK027471.1 | 7 | 841 | BAB55136.1 | |
| AK027501.1 | 10 | 1199 | BAB55158.1 | |
| AK027654.1 | 7 | 848 | BAB55269.1 | |
| AK027867.1 | 17 | 2015 | BAB55418.1 | |
| AK074975.1 | 7 | 759 | BAC11326.1 | |
| AK096337.1 | 2 | 2176 | ||
| AK302928.1 | 2 | 544 | ||
| AK315143.1 | 20 | 2307 |
Target Gene Details
Entrez Gene ID: | 101927233 |
Gene Name: | SEMA6A antisense RNA 1 |
Gene Aliases: |
CTB-118N6.3 |
Location: |
Chr.5:116447498-116471673 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6A-AS1 | NR_109879.1 | 1 | 101 | |
| CD300011.1 |
More Information
Set Membership: |
|

Genomic Map