Assay Details
Target Gene Details
Entrez Gene ID: | 4155 |
Gene Name: | myelin basic protein |
Gene Aliases: |
- |
Location: |
Chr.18:76978833-77133708 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBP | NM_001025081.1 | 7 | 1388 | NP_001020252.1 |
| NM_001025090.1 | 6 | 1310 | NP_001020261.1 | |
| NM_001025092.1 | 5 | 1277 | NP_001020263.1 | |
| NM_001025101.1 | 9 | 1882 | NP_001020272.1 | |
| NM_002385.2 | 6 | 1355 | NP_002376.1 | |
| AK094611.1 | 1 | 1474 | ||
| AK095121.1 | 1 | 2264 | ||
| AK098402.1 | 6 | 1266 | ||
| AK122594.1 | 2 | 1833 | ||
| AK123433.1 | 2 | 3930 | BAC85616.1 | |
| AK124830.1 | 2 | 2607 | BAC85961.1 | |
| AK126858.1 | 1 | 4424 | ||
| AK128770.1 | 6 | 1269 | ||
| AK128788.1 | 6 | 1266 | ||
| AK289893.1 | ||||
| AK293922.1 | ||||
| BC008749.2 | 6 | 1248 | ||
| BC080654.1 | 5 | 1222 | AAH80654.1 | |
| CR627018.1 | 4 | 9040 | CAH10359.1 | |
| M13577.1 | 6 | 1255 | AAA59562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
|
| esv3643197 | Chr.18:76911767 - 77002929 on Build GRCh38 | Loss |
|
| nsv1063632 | Chr.18:76317786 - 77170410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map