Assay Details
Target Gene Details
Entrez Gene ID: | 23357 |
Gene Name: | angel homolog 1 |
Gene Aliases: |
Ccr4e, KIAA0759 |
Location: |
Chr.14:76786473-76812947 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGEL1 | NM_015305.3 | NP_056120.2 | ||
| XM_017021115.1 | XP_016876604.1 | |||
| AB018302.1 | ||||
| AK301069.1 | ||||
| AK301589.1 | ||||
| AL137268.1 | CAB70667.2 | |||
| BC028714.2 | AAH28714.1 |
More Information
Set Membership: |
|

Genomic Map