Assay Details
Target Gene Details
Entrez Gene ID: | 222545 |
Gene Name: | G protein-coupled receptor class C group 6 member A |
Gene Aliases: |
GPCR, bA86F4.3 |
Location: |
Chr.6:116788512-116831128 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRC6A | NM_001286354.1 | NP_001273283.1 | ||
| NM_001286355.1 | 3 | 1295 | NP_001273284.1 | |
| NM_148963.3 | 3 | 1295 | NP_683766.2 | |
| XM_017010475.1 | 4 | 3228 | XP_016865964.1 | |
| XM_017010476.1 | 3 | 1256 | XP_016865965.1 | |
| AF502962.1 | 3 | 1273 | AAM22230.1 | |
| AY435125.1 | 3 | 1284 | AAS13465.1 | |
| AY435126.1 | AAS13466.1 | |||
| AY435127.1 | 3 | 1284 | AAS13467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map