Assay Details
Target Gene Details
Entrez Gene ID: | 64327 |
Gene Name: | limb development membrane protein 1 |
Gene Aliases: |
ACHP, C7orf2, DIF14, LSS, PPD2, THYP, TPT, ZRS |
Location: |
Chr.7:156673354-156893208 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBR1 |
Target Gene Details
Entrez Gene ID: | 140545 |
Gene Name: | ring finger protein 32 |
Gene Aliases: |
FKSG33, HSD15, LMBR2 |
Location: |
Chr.7:156640286-156678333 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF32 | NM_001184996.1 | 9 | 1766 | NP_001171925.1 |
| NM_001184997.1 | 9 | 1619 | NP_001171926.1 | |
| NM_001308273.1 | 9 | 1841 | NP_001295202.1 | |
| NM_030936.3 | 9 | 1661 | NP_112198.1 | |
| NR_131778.1 | 9 | 1615 | ||
| XM_005249522.4 | 9 | 1573 | XP_005249579.1 | |
| XM_011515804.2 | 10 | 1683 | XP_011514106.1 | |
| XM_011515805.2 | 10 | 1840 | XP_011514107.1 | |
| XM_011515806.2 | 9 | 1751 | XP_011514108.1 | |
| XM_011515807.2 | 9 | 1559 | XP_011514109.1 | |
| XM_011515808.2 | 9 | 1671 | XP_011514110.1 | |
| XM_011515809.2 | 9 | 1744 | XP_011514111.1 | |
| XM_011515810.2 | 9 | 1561 | XP_011514112.1 | |
| XM_011515811.2 | 9 | 1556 | XP_011514113.1 | |
| XM_011515812.2 | 9 | 1663 | XP_011514114.1 | |
| XM_011515813.2 | 9 | 1622 | XP_011514115.1 | |
| XM_017011753.1 | XP_016867242.1 | |||
| AF325690.1 | 9 | 1569 | AAG50281.1 | |
| AF441222.1 | 8 | 1508 | AAM18664.1 | |
| AF441224.1 | 9 | 1615 | AAM18666.1 | |
| AL136874.1 | 9 | 1549 | CAB66808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv609209 | Chr.7:156663585 - 156716626 on Build GRCh38 | Gain |
|
| nsv6020 | Chr.7:156657458 - 156687987 on Build GRCh38 | Insertion |
|
| nsv831198 | Chr.7:156629947 - 156829662 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map