Assay Details
Target Gene Details
Entrez Gene ID: | 89872 |
Gene Name: | aquaporin 10 |
Gene Aliases: |
AQPA_HUMAN |
Location: |
Chr.1:154321059-154325325 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AQP10 | NM_080429.2 | 6 | 1708 | NP_536354.2 |
| XM_011510104.2 | 6 | 1768 | XP_011508406.1 | |
| AB066105.1 | 6 | 1708 | BAB91223.1 | |
| AF159174.1 | 5 | 2179 | AAL25998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523934 | Chr.1:154323600 - 154335154 on Build GRCh38 | Loss |
|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map