Assay Details
Target Gene Details
Entrez Gene ID: | 115811 |
Gene Name: | IQ motif containing D |
Gene Aliases: |
4933433C09Rik, CFAP84, DRC10 |
Location: |
Chr.12:113195441-113221344 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCD | NM_138451.2 | 2 | 482 | NP_612460.1 |
| XM_005253832.4 | 2 | 725 | XP_005253889.1 | |
| XM_005253833.4 | 2 | 973 | XP_005253890.1 | |
| XM_005253834.4 | XP_005253891.1 | |||
| XM_011537862.2 | 2 | 1605 | XP_011536164.1 | |
| XM_011537863.2 | 3 | 851 | XP_011536165.1 | |
| XM_011537864.1 | 2 | 482 | XP_011536166.1 | |
| XM_011537865.2 | XP_011536167.1 | |||
| XM_011537866.2 | XP_011536168.1 | |||
| XM_017018771.1 | XP_016874260.1 | |||
| AK127152.1 | 2 | 482 | BAC86856.1 | |
| BC013151.1 | 2 | 463 | AAH13151.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|

Genomic Map